Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-azidobenzyl alcohol | 31499-54-4 | MFCD19439656 | 99.9% | 149.15 g/mol | C7H7N3O | 10 MG
SDP

Supplier:  Medchemexpress LLC HY15184010MG

Encompass_Preferred

4-Azidobenzyl alcohol is an aryl azide reagent used in click chemistry and bioconjugation. It provides an azide functional group and a primary hydroxymethyl handle that enable copper-catalyzed and strain-promoted azide-alkyne cycloadditions for introducing azide functionality into molecules.

  • High purity (99.9%).
  • Suitable for CuAAC and SPAAC reactions.
  • Contains an azide group and a hydroxymethyl reactive handle.
  • Soluble in DMSO (≈250 mg/mL; ultrasonic assistance recommended).
  • Molecular formula C7H7N3O; molecular weight 149.15 g/mol.
  • Recommended storage: stock solution -80 °C (6 months) or -20 °C (1 month) under nitrogen.

Catalog No. 50-002-87662


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.