Learn More
Medchemexpress LLC 4-Azidobenzyl alcohol | 31499-54-4 | MFCD19439656 | C7H7N3O | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY151840
Encompass_Preferred
4-Azidobenzyl alcohol is a click chemistry reagent featuring an azide group. It is designed to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. This reagent facilitates rapid synthesis of new compounds through selective, modular, wide-ranging, and high-yield chemical reactions, making it ideal for various chemical and bioorthogonal applications.
  • Click chemistry reagent with an azide group
  • Participates in CuAAc reactions with alkyne groups
  • Undergoes SPAAC reactions with DBCO or BCN groups
  • High purity of 99.90%
  • Appearance as a light yellow to yellow solid
  • For research use only
Catalog No. 50-003-74544