Learn More
Medchemexpress LLC 4-(Benzyloxy)butanal | 5470-84-8 | MFCD11553544 | 178.23 g/mol | C11H14O2 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW1432301G
Encompass_Preferred
4-(Benzyloxy)butanal is a small-molecule linker containing an acid-labile benzyl protecting group and a reactive primary aldehyde. It is used in bioconjugation and probe synthesis, where the aldehyde reacts with hydrazide and aminooxy groups to form hydrazones or oximes for labeling or crosslinking. Molecular formula C11H14O2; molecular weight 178.23 g/mol. Available in multiple small-scale package sizes for research use.
  • Reactive primary aldehyde for hydrazone and oxime formation.
  • Acid-labile benzyl protecting group enables controlled deprotection.
  • Suitable for labeling and crosslinking oxidized carbohydrates.
  • Useful as a linker in probe and conjugate synthesis.
  • Available in multiple small-scale package sizes for laboratory research.
Catalog No. 50-002-83492