Learn More
Medchemexpress LLC 4-(Benzyloxy)butanal | 5470-84-8 | MFCD11553544 | 97.0% | 178.23 g/mol | C11H14O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW143230100MG
Encompass_Preferred
4-(Benzyloxy)butanal is a benzyl-protected primary aldehyde used as a linker and synthetic intermediate for bioconjugation and organic synthesis. The benzyl protecting group is acid-labile, while the terminal aldehyde reacts selectively with hydrazide and aminooxy nucleophiles, enabling efficient labeling and crosslinking of carbonyl-containing biomolecules such as oxidized carbohydrates.
  • Benzyl-protected primary aldehyde.
  • Acid-labile protecting group for controlled deprotection.
  • Reactive toward hydrazide and aminooxy groups for efficient bioconjugation.
  • Suitable for labeling and crosslinking oxidized carbohydrates.
  • Storage recommended under nitrogen: 4°C (neat); in solvent -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-002-83037