Learn More
Medchemexpress LLC 4-Boc-2-oxopiperazine | 76003-29-7 | 200.23 | 500 G

Supplier: Medchemexpress LLC HYW001639500G
4-Boc-2-Oxopiperazine is a biochemical reagent primarily used as a biological material or organic compound for various life science research applications.
- Purity of 98.0%
- Appears as a white to off-white solid
- Molecular formula C9H16N2O3
- Soluble in DMSO at 25 mg/mL
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.