Learn More
Medchemexpress LLC 4-Boronobenzoic acid | 14047-29-1 | 99.9% | C7H7BO4 | 500 G
SDP
Supplier:  Medchemexpress LLC HY75102500G
Encompass_Preferred
4-Boronobenzoic acid is an organic compound. It forms reversible cyclic boronate esters with molecules containing cis-vicinal diol structures, such as sialic acid, through the boron atom, allowing for molecular recognition and binding.
  • Can be used in breast cancer research.
  • Forms reversible cyclic boronate esters.
  • Allows for molecular recognition and binding.
Catalog No. 50-004-34820