Learn More
Medchemexpress LLC 4-Bromo-1,1'-biphenyl-d9 | 142475-00-1 | 242.16 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY41826S1
Encompass_Preferred
4-Bromo-1,1'-biphenyl-d9 is a deuterium labeled compound of 4-Bromo-1,1'-biphenyl, intended for research use only. It can be used as a tracer and as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Suitable for research applications
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Applicable with NMR, GC-MS, or LC-MS techniques
Catalog No. 50-004-03173