Learn More
Medchemexpress LLC 4-bromo-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione | 2093536-12-8 | MFCD31730866 | 99.7% | 337.13 g/mol | C13H9BrN2O4 | 25 MG

Supplier: Medchemexpress LLC 25MGHYW039116
Thalidomide-4-Br is a brominated derivative of thalidomide used as a research compound for studies in inflammation, oncology, and neurology. The compound is supplied as an off-white to gray solid with confirmed identity by 1H NMR and LCMS and a high HPLC purity suitable for research applications.
- Chemical formula: C13H9BrN2O4.
- Molecular weight: 337.13 g/mol.
- CAS number: 2093536-12-8.
- Purity (HPLC): 99.7%.
- Appearance: off-white to gray solid.
- Identity confirmed by 1H NMR and LCMS.
- Storage (powder): -20°C for up to 3 years, 4°C for 2 years.
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month.
- Application: research in inflammation, oncology, and neurology.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.