Learn More
Medchemexpress LLC 4-Bromo-2-methoxyphenol | 7368-78-7 | 203.03 | 5 G

Supplier: Medchemexpress LLC HYW0078655G
4-Bromo-2-methoxyphenol is a pharmaceutical intermediate that can be used for the synthesis of 2-(o-Tolyloxy)-5-hexylphenol (PT-70).
- Appearance: <34°C solid, >37°C liquid
- Color: Off-white to yellow
- Solubility: 200 mg/mL in DMSO (985.08 mM)
- Storage (pure form): -20°C for 3 years, 4°C for 2 years
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.