Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (4-bromo-2-(methylamino)phenyl)diethylphosphine oxide | 2685825-06-1 | MFCD35092689 | 99.8% | 290.14 g/mol | C11H17BrNOP | 100 G
SDP

Supplier:  Medchemexpress LLC HYW441592100G

Encompass_Preferred

(4-Bromo-2-(methylamino)phenyl)diethylphosphine oxide is a brominated aryl diethylphosphine oxide used as a drug intermediate in the synthesis of active pharmaceutical compounds. It is supplied as a solid with high analytical purity and is characterized by COA and SDS data for research and chemical synthesis applications.

  • High purity: 99.8% (LCMS).
  • Identified by CAS 2685825-06-1.
  • Chemical formula C11H17BrNOP, molecular weight 290.14 g/mol.
  • Appearance: light brown to brown solid.
  • Low water content (KF 0.06%).
  • Storage: stable at room temperature for up to 3 years; in solvent store at -80°C or -20°C as specified.
  • Intended use: reagent for organic synthesis and drug intermediate applications.

Catalog No. 50-002-81506


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.