Learn More
Medchemexpress LLC 4-Bromo-2-methylpyridine | 22282-99-1 | 172.02 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW007447
Encompass_Preferred
4-Bromo-2-methylpyridine is a colorless to light yellow liquid biochemical reagent, useful as a biological material or organic compound for life science research.
  • Used as a starting material in the preparation of crown-ester-bipyridines and viologens through sodium or nickel reactive coupling, side chain oxidation and esterification.
  • Serves as an important raw material and intermediate in organic synthesis, pharmaceuticals, agrochemicals, and in dyes.
Catalog No. 50-004-49295