Learn More
Accela Chembio Inc 4-bromo-3-fluorobenzaldehyde | 25g | 133059-43-5 | MFCD03095000 | 97+% | Shelf Life: 1260 Days | Light Sensitive/air Sensitive/+4
Supplier:  Accela Chembio Inc SY01436225G
Encompass_Preferred
4-bromo-3-fluorobenzaldehyde | 25g | 133059-43-5 | MFCD03095000 | 97+% | Shelf Life: 1260 Days | Light Sensitive/air Sensitive/+4
Catalog No. 50-251-987