Learn More
Medchemexpress LLC 4-Bromo-3-fluorobenzaldehyde | 133059-43-5 | 203.01 | 5 G
SDP
Supplier:  Medchemexpress LLC 25GHYW007518
Encompass_Preferred
4-Bromo-3-fluorobenzaldehyde is a drug intermediate that can be used for the synthesis of triaryl dimer antibacterial compounds.
  • For research use only.
  • Not for sale to patients.
Catalog No. 50-004-49181