Learn More
Medchemexpress LLC HY-W003445 100mg Medchemexpress, 4-Bromo-3-hydroxybenzoic acid CAS:14348-38-0 Purity:>98%
Supplier: Medchemexpress LLC HYW003445100MG
Medchemexpress, HY-W003445 100mg 4-Bromo-3-hydroxybenzoic acid CAS:14348-38-0 4-Bromo-3-hydroxybenzoic acid is a metabolite of Brocresine and a histidine decarboxylase (HDC) inhibitor with IC50s of 1 mM for both rat fetal and rat gastric HDC. 4-Bromo-3-hydroxybenzoic acid also inhibits aromatic-L-amino acid decarboxylase from hog kidney and rat gastric mucosa in vitro with IC50s of 1 mM for both enzymes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.