Learn More
Medchemexpress LLC 4-bromo-6-[(3,4-dichlorophenyl)sulfanyl]-1-{[4-(dimethylcarbamoyl)phenyl]methyl}-1H-indole-2-carboxylic acid | 2216763-38-9 | MFCD31714165 | 99.0% | 578.30 | C25H19BrCl2N2O3S | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12479810MM 1ML
Encompass_Preferred
Rheb inhibitor NR1 is a small-molecule probe that binds Rheb in the switch II domain and selectively inhibits mTORC1 signaling. It is provided in solution and solid formats for use in biochemical and cellular research, with characterization data and batch-level purity available for quality assessment.
  • Selective inhibitor of Rheb that blocks mTORC1 signaling.
  • Binds the switch II domain of Rheb.
  • Supplied as a 10 mM solution in DMSO (1 mL) and as solids in multiple mass sizes.
  • High purity suitable for research (≈99.0%).
  • Molecular formula C25H19BrCl2N2O3S; molecular weight 578.30.
  • Intended for use as a chemical probe in cellular and biochemical assays.
Catalog No. 50-002-39669