Learn More
Medchemexpress LLC 4-Bromo-7-azaindole | 348640-06-2 | MFCD08272233 | 97% | 197.03 | 5 G
SDP
Supplier:  Medchemexpress LLC HY300145G
Encompass_Preferred
4-Bromo-7-azaindole is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also known as 4-bromo-1h-pyrrolo[2,3-b]pyridine and appears as a light yellow solid.
  • Used as a biological material or organic compound for life science related research
  • Categorized as a building block
  • Appears as a light yellow solid
  • Should be stored at room temperature
Catalog No. 50-004-04812