Learn More
Medchemexpress LLC 4'-bromo-resveratrol | 1224713-90-9 | 99.9% | 291.14 g/mol | C14H11BrO2 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12411310MM 1ML
Encompass_Preferred
4'-Bromo-resveratrol is a brominated derivative of resveratrol that functions as a dual inhibitor of sirtuin-1 and sirtuin-3. Supplied as a 10 mM solution in DMSO (1 mL), it is intended for in vitro biochemical and cell-based research focused on sirtuin signaling, apoptosis, and metabolic regulation.
  • Ready-to-use 10 mM solution in DMSO.
  • Suitable for in vitro enzyme inhibition assays and cell-based studies.
  • Acts as a dual SIRT1 and SIRT3 inhibitor.
  • High purity (≈99.9%) for reproducible results.
  • Storage: in solvent -80°C (6 months) or -20°C (1 month); protect from light.
Catalog No. 50-002-38421