Learn More
Medchemexpress LLC 4'-Bromoacetophenone | 99-90-1 | 100.0% | 199.04 | 50 G
SDP
Supplier:  Medchemexpress LLC HY2043650G
Encompass_Preferred
4-Bromoacetophenone is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It serves as a fundamental starting material for organic synthesis.
  • Acts as a biochemical reagent
  • Can be used as a biological material or organic compound
  • Suitable for life science related research
  • Possesses activity against positive phototaxis of Chlamydomonas cells
  • Fundamental starting material for organic synthesis
Catalog No. 50-004-01890