Learn More
Medchemexpress LLC 4-Bromobenzaldehyde | 1122-91-4 | 185.02 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW001138
Encompass_Preferred
4-Bromobenzaldehyde is a versatile biochemical reagent suitable for use as a biological material or organic compound in life science research. It is widely utilized as an intermediate in the preparation of agrochemicals, pharmaceuticals, and various other organic compounds.
  • Precursor for Schiff's base, enhancing PVC photostability
  • Plays a role in palladium-catalyzed mono- and bis-arylation of 3,4-(ethylenedioxy)thiophenes
  • Employed in cross-coupling studies with potassium vinyltrifluoroborate
Catalog No. 50-004-49998