Learn More
AdipoGen 4-Bromobenzenediazonium BF4
SDP
Supplier:  AdipoGen CDXB0531G005
Encompass_Preferred
Chemical. CAS 673-40-5. Formula BrC6H4N2 . BF4. MW 270.82. Building block and intermediate used in synthesis and chemical functionalization of fluorescent surfaces.
Catalog No. 50-256-1587