Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Bromobenzoic acid | 586-76-5 | 99.9% | C7H5BrO2 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHY78700

Encompass_Preferred

4-Bromobenzoic acid is a tyrosinase inhibitor, with an IC50 range of 0-0.3 mg/mL. This compound is metabolized via the protocatechuic acid (PCA) pathway and is suitable for research on cosmetic whitening or pigmentation conditions.

  • Tyrosinase inhibitor with an IC50 range of 0-0.3 mg/mL
  • Metabolized via the protocatechuic acid (PCA) pathway
  • Suitable for research on cosmetic whitening
  • Suitable for research on pigmentation conditions
  • Molecular weight: 201.02
  • Appearance: Solid, white to off-white
  • Purity: 99.9%
  • Storage conditions: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-59118


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.