Learn More
Medchemexpress LLC 4-bromobenzoic acid-d4 | 787624-24-2 | MFCD06658868 | 98.1% | 205.04 g/mol | C7HD4BrO2 | 1 G

Supplier: Medchemexpress LLC HYW2492571G
4-Bromobenzoic acid-d4 is a deuterium-labeled analogue of 4-bromobenzoic acid intended for use as a tracer and internal standard in analytical workflows. The compound is supplied as both solid material and as a solution format and is characterized for known isotopic labeling to support quantitative analyses in NMR, GC-MS, and LC-MS.
- Deuterium-labeled analogue for use as an internal standard
- Suitable for quantitative NMR, GC-MS, and LC-MS analyses
- Molecular weight 205.04 g/mol
- High purity (98.1%) for analytical applications
- Available as solid and as 10 mM solution in DMSO
- CAS number 787624-24-2 for unambiguous identification
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.