Learn More
Medchemexpress LLC 4 -Bromoflavone 10mM 1mL | 20525-20-6 | 301.13 g·mol⁻¹ | C15H9BrO2 | 1 ML

Supplier: Medchemexpress LLC HY11577210MM 1ML
4′-Bromoflavone (CAS 20525-20-6) is a flavone derivative used as a research reagent and a potent inducer of phase II detoxification enzymes. Supplied as a ready-to-use 10 mM solution in DMSO, it is intended for in vitro studies of chemoprevention and enzyme regulation.
- Provided as 10 mM solution in DMSO, 1 mL volume.
- High purity (≈97.7%) suitable for research applications.
- Molecular weight 301.13 g·mol⁻¹; formula C15H9BrO2.
- Useful for studies of phase II enzyme induction and chemopreventive mechanisms.
- Product documentation and safety data sheet are available from the supplier.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.