Learn More
eMolecules​ 4-Bromotoluene | Oakwood Chemical | 106-38-7 | MFCD00000109 | 171.037 | C7H7Br | 98.000 | Cc1ccc(Br)cc1 | 100g | 537661865
Supplier:  eMolecules​ 537661865
Encompass_Preferred
4-Bromotoluene | Oakwood Chemical | 106-38-7 | MFCD00000109 | 171.037 | C7H7Br | 98.000 | Cc1ccc(Br)cc1 | 100g | 537661865
Catalog No. NC3724077