Learn More
Medchemexpress LLC 4-(Butylamino)benzoic acid-d7 | 1219803-44-7 | 99.7% | 200.29 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY41463S5MG
Encompass_Preferred
4-(Butylamino)benzoic acid-d7 is a deuterium labeled compound that can be used as a tracer and an internal standard for quantitative analysis. Deuteration, the incorporation of stable heavy isotopes, has shown potential in affecting the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-04877