Learn More
Medchemexpress LLC 4-butylphenol-d5 | 1219795-04-6 | 98.0% | 155.25 g·mol^-1 | C10H9D5O | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW144683S10MG
Encompass_Preferred
4-Butylphenol-d5 is a deuterium-labeled analogue of 4-butylphenol used as an isotopically labeled standard or tracer in analytical and research applications. It is supplied in small, research-scale quantities for laboratory use and intended for scientific research only.
  • Deuterium-labeled compound suitable as an internal standard in mass spectrometry.
  • High reported purity for reliable analytical performance.
  • Molecular formula provided to support quantitation and identification.
  • Available in small, convenient pack sizes for research workflows.
Catalog No. 50-002-83740