Learn More
Medchemexpress LLC 4-butylphenol-d5 | 1219795-04-6 | 98.3% | 155.25 g/mol | C10H9D5O | 5 MG

Supplier: Medchemexpress LLC HYW144683S5MG
4-Butylphenol-d5 is the deuterium-labeled analogue of 4-butylphenol intended for use as a tracer and internal standard in quantitative analytical workflows such as NMR, GC-MS, and LC-MS.
- Deuterium-labeled internal standard for quantitative analysis.
- Suitable for NMR, GC-MS, and LC-MS applications.
- Molecular formula C10H9D5O.
- Molecular weight 155.25 g/mol.
- CAS number 1219795-04-6.
- Available in small-mass vials (1 mg, 5 mg, 10 mg).
- Typical purity reported ~98.3% by distributors.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.