Learn More
Cayman Chemical 4-carboxy TEMPO 500mg
SDP
Supplier:  Cayman Chemical 23139500 MG
Encompass_Preferred
A spin label; reacts with ROS to monitor ROS production in vivo using ESR spectroscopy
Catalog No. 50-299-3956