Learn More
Medchemexpress LLC 4-Carboxypyrazole (1H-Pyrazole-4-carboxylic acid) | 37718-11-9 | 112.09 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW002065100G
Encompass_Preferred
4-Carboxypyrazole (1H-Pyrazole-4-carboxylic acid) is a metabolite of Fomepizole. It is involved in the synthesis of ALKBH1 inhibitors and can be used in gastric cancer research.
  • Metabolite of Fomepizole.
  • Involved in the synthesis of ALKBH1 inhibitors.
  • Can be used in gastric cancer research.
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-004-47520