Learn More
Medchemexpress LLC 4-Carboxypyrazole | 37718-11-9 | 112.09 | 1 ML

Supplier: Medchemexpress LLC HYW00206510MM/1ML
4-Carboxypyrazole (1H-Pyrazole-4-carboxylic acid) is a metabolite of Fomepizole. It is involved in the synthesis of ALKBH1 inhibitors and can be used in the research of gastric cancer.
- Metabolite of Fomepizole.
- Involved in the synthesis of ALKBH1 inhibitors.
- Can be used in gastric cancer research.
- Molecular formula: C4H4N2O2.
- Appearance: White to off-white solid.
- Powder storage: -20°C for 3 years, 4°C for 2 years.
- In solvent storage: -80°C for 6 months, -20°C for 1 month.
- Soluble in DMSO (100 mg/mL).
- For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.