Learn More
Medchemexpress LLC 4-Carboxypyrazole | 37718-11-9 | 112.09 | 10 G

Supplier: Medchemexpress LLC HYW00206510G
4-Carboxypyrazole (1H-Pyrazole-4-carboxylic acid) is a metabolite of Fomepizole. It is involved in the synthesis of ALKBH1 inhibitors and can be used in the research of gastric cancer.
- Solid appearance
- White to off-white color
- Derived from endogenous metabolite
- Powder storage: -20°C for 3 years; 4°C for 2 years
- In solvent storage: -80°C for 6 months; -20°C for 1 month
- Functions as a drug metabolite or drug intermediate
- Involves metabolic enzyme/protease pathways
- In vitro solubility in DMSO: 100 mg/mL (892.16 mM)
- Soluble in various in vivo dissolution protocols
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.