Learn More
Medchemexpress LLC 4-Carboxypyrazole | 37718-11-9 | 112.09 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00206550G
Encompass_Preferred
4-Carboxypyrazole (1H-Pyrazole-4-carboxylic acid) is a metabolite of Fomepizole. It is involved in the synthesis of ALKBH1 inhibitors and can be used in the research of gastric cancer. The appearance of the product is a white to off-white solid. It is classified as Ketones, Aldehydes, and Acids, and its initial source is an endogenous metabolite. For research use only.
  • White to off-white solid
  • Involved in the synthesis of ALKBH1 inhibitors
  • Can be used in the research of gastric cancer
  • Classified as ketones, aldehydes, and acids
  • Initial source is an endogenous metabolite
  • For research use only
Catalog No. 50-004-48839