Learn More
Medchemexpress LLC 4-Carboxypyrazole (Standard) | 37718-11-9 | 112.09 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0020655G
Encompass_Preferred
4-Carboxypyrazole is an endogenous metabolite. The compound is of analytical standard grade and serves as a reference standard for assays. It is commonly utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Endogenous metabolite.
  • Analytical standard grade.
  • Reference standard for assays.
  • Suitable for qualitative, quantitative, and methodological research in HPLC, GC, and MS.
Catalog No. 50-004-47484