Learn More
Bellco Glass, Inc. 4 Chamber Cell Culture Slide
SDP
Supplier:  Bellco Glass, Inc. 229164000
Encompass_Preferred
Grow cells directly on the slide without any need for future transfer; ideal for growing and viewing cells under a microscope on standard 25mm x 75mm slideIntegrated gasket prevents cross-contamination between chambersInert hydrophobic green border printed on slide to define cell culture areaChamber holder features easy flip locks for assembly/disassembly; no separation tool requiredPacking trays can be used as incubation racks in CO2 incubatorsUniform hydrophilic surface ideal for consistent cell attachmentSlides are labeled with numeric markingsAdhesive freeNot recommended for use with acetone Pack Description6/Tray with 2 Trays/BoxCase Quantity12SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialSlide: GlassSecondary MaterialChambers: PolystyreneWorking Volume0.5-1.3mLTissue Culture TreatedNoCell Growth Area2.13cm2RNase/DNase FreeYes
Catalog No. 50-233-0811