Learn More
Medchemexpress LLC 4-chloro-2-methoxyphenol | 16766-30-6 | MFCD00070774 | 99.9% | 158.58 | C7H7ClO2 | 5 G

Supplier: Medchemexpress LLC HYW0391695G
4-Chloroguaiacol (4-chloro-2-methoxyphenol) is a chlorinated guaiacol derivative used as a research reagent. It has reported antimicrobial activity against Staphylococcus aureus and Escherichia coli (MIC 110 μg/mL) and is supplied as a high-purity material for laboratory studies.
- Chemical name: 4-chloro-2-methoxyphenol
- CAS number: 16766-30-6
- Chemical formula: C7H7ClO2
- Molecular weight: 158.58 g/mol
- Purity: 99.89%
- Appearance: colorless to off-white; solid below ~16°C
- Storage: keep at 4°C, store under nitrogen; in solvent keep frozen for long-term storage
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.