Learn More
Medchemexpress LLC 4-Chloro-2-methylphenol-d4 | 1219804-06-4 | 99.6% | 146.61 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY34498S5MG
Encompass_Preferred
4-Chloro-2-methylphenol-d4 is the deuterium labeled 4-Chloro-2-methylphenol. It is used for research purposes, such as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-03725