Learn More
Medchemexpress LLC 4-chloro-3-methylisoxazol-5-amine | 166964-09-6 | 99.8% | C4H5ClN2O | 50 G
SDP
Supplier:  Medchemexpress LLC HY7006550G
Encompass_Preferred
4-Chloro-3-methylisoxazol-5-amine (5-Amino-4-chloro-3-methylisoxazole) is a drug intermediate used for the synthesis of various active compounds. For storage, keep at room temperature (RT) and protect from light. In solvent, store at -80°C for 1 year, or -20°C for 6 months.
  • Drug intermediate for the synthesis of various active compounds.
Catalog No. 50-004-34363