Learn More
Medchemexpress LLC 4-Chloro-3-nitrobenzoic acid | 96-99-1 | 99.4% | 201.56 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHY20559
Encompass_Preferred
4-Chloro-3-nitrobenzoic acid has oral activity, and oral administration can affect the synthesis activity of liver proteins and the activities of urokinase and histidine kinase. It is intended for research use only.
  • High purity (99.39%)
  • Solid appearance, white to off-white color
  • Stable under recommended storage conditions
  • Soluble in DMSO for in vitro experiments
  • Multiple in vivo dissolution protocols available
  • Suitable for laboratory chemicals and substance manufacturing applications
Catalog No. 50-004-03219