Learn More
Medchemexpress LLC 4-Chloro-3-nitrobenzoic acid | 96-99-1 | 99.4% | 201.56 | 500 G
SDP
Supplier:  Medchemexpress LLC HY20559500G
Encompass_Preferred
4-Chloro-3-nitrobenzoic acid has oral activity. Oral administration of this compound can affect the synthesis activity of liver proteins and the activities of urokinase and histidine kinase. It is intended for research use only.
  • Has oral activity.
  • Affects the synthesis activity of liver proteins.
  • Affects the activities of urokinase and histidine kinase.
Catalog No. 50-004-01211