Learn More
Medchemexpress LLC 4-Chloro-3-sulfamoylbenzoic acid | 1205-30-7 | 235.64 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01216850G
Encompass_Preferred
4-Chloro-3-sulfamoylbenzoic acid is an inhibitor of human carbonic anhydrase, targeting the cytosolic isoforms hCA I and hCA II (glaucoma-associated) and the transmembrane isoforms hCA IV and hCA IX (tumor hypoxia-induced). This compound is intended for research applications.
  • Inhibits human carbonic anhydrase
  • Targets cytosolic isoforms hCA I and hCA II
  • Targets transmembrane isoforms hCA IV and hCA IX
  • Stable under recommended storage conditions
  • Appears as a solid
  • Melting point: 256-258 °C
Catalog No. 50-004-53006