Learn More
Medchemexpress LLC 4-Chlorobenzoic acid | 74-11-3 | 99% | 156.57 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHY41465
Encompass_Preferred
4-Chlorobenzoic acid (Standard) is the analytical standard of 4-Chlorobenzoic acid (HY-41465). This product is intended for research and analytical applications. 4-Chlorobenzoic acid is a halogenated aromatic compound that can be used as an industrial chemical and an intermediate in drug synthesis. 4-Chlorobenzoic acid is a persistent organic micropollutant and can be biodegraded through a direct dehalogenation pathway.
  • Intended for research and analytical applications
  • Used as an industrial chemical and an intermediate in drug synthesis
  • Biodegraded through a direct dehalogenation pathway
Catalog No. 50-004-02476