Learn More
Medchemexpress LLC 4-Chlorobenzoic acid | 74-11-3 | 99.37% | 156.57 | 50 G
SDP
Supplier:  Medchemexpress LLC HY4146550G
Encompass_Preferred
4-Chlorobenzoic acid is a halogenated aromatic compound used as an industrial chemical and an intermediate in drug synthesis. It is also a persistent organic micropollutant that can be biodegraded through a direct dehalogenation pathway.
  • Halogenated aromatic compound.
  • Used as an industrial chemical.
  • Intermediate in drug synthesis.
  • Persistent organic micropollutant.
  • Biodegraded through a direct dehalogenation pathway.
Catalog No. 50-004-04656