Learn More
Medchemexpress LLC 4-Chlorobenzoic acid | 74-11-3 | 99.4% | 156.57 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY41465
Encompass_Preferred
4-Chlorobenzoic acid is a halogenated aromatic compound used as an industrial chemical and an intermediate in drug synthesis. It is a persistent organic micropollutant that can be biodegraded through a direct dehalogenation pathway.
  • Used as an industrial chemical
  • Intermediate in drug synthesis
  • Persistent organic micropollutant
  • Biodegraded through a direct dehalogenation pathway
  • Suitable for biochemical assays
  • Suitable for cell assays
Catalog No. 50-004-03033