Learn More
Medchemexpress LLC 4-Chlorobenzoic acid | 74-11-3 | MFCD00002531 | 99.4% | 156.57 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY4146510MM/1ML
Encompass_Preferred
4-Chlorobenzoic acid is a halogenated aromatic compound that can be used as an industrial chemical and an intermediate in drug synthesis. It is a persistent organic micropollutant and can be biodegraded through a direct dehalogenation pathway.
  • Halogenated aromatic compound
  • Used as an industrial chemical
  • Intermediate in drug synthesis
  • Persistent organic micropollutant
  • Can be biodegraded through a direct dehalogenation pathway
Catalog No. 50-004-04515