Learn More
Medchemexpress LLC 4-Chlorophenylacetic acid | 1878-66-6 | 99.97% | 170.59 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW010062500G
Encompass_Preferred
4-Chlorophenylacetic acid is a halogenated phenylacetic acid derivative that acts as a potent aromatase inhibitor and antagonizes estrogen signaling. It can also serve as a carbon and energy source for *Pseudomonas* sp. strain CBS3. It is also effective against estrogen-induced mammary tumorigenesis.
  • Potent aromatase inhibitor.
  • Antagonizes estrogen signaling.
  • Can serve as a carbon and energy source for *Pseudomonas* sp. strain CBS3.
  • Effective against estrogen-induced mammary tumorigenesis.
Catalog No. 50-004-52543