Learn More
Medchemexpress LLC 4-chloroquinolin-8-ol | 57334-36-8 | MFCD00661657 | 99.0% | 179.61 g·mol⁻¹ | C9H6ClNO | 5 G
SDP
Supplier:  Medchemexpress LLC HYW2427095G
Encompass_Preferred
4-Chloroquinolin-8-ol is a chlorinated quinolinol used as an Alq3-derived aluminum ligand in organic electronics research. It is employed to prepare aluminum coordination complexes for studies of electron transport and emissive layers in OLEDs, and is supplied as a high-purity reagent for materials and optoelectronic investigations.
  • High chemical purity suitable for research applications.
  • Useful as a ligand for Alq3-type aluminum complexes.
  • Supplied in multiple package sizes to suit experimental needs.
  • Stable when stored at 4°C under nitrogen; solvent storage recommendations available.
  • Well characterized with CAS number 57334-36-8 and known molecular weight.
Catalog No. 50-002-80488