Learn More
Medchemexpress LLC 4-chlorosalicylic acid | 5106-98-9 | MFCD00002449 | 100.0% | 172.57 | C7H5ClO3 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0168675G
Encompass_Preferred
4-Chlorosalicylic acid is a chlorinated salicylic (benzoic acid) derivative used as a pharmaceutical intermediate and research reagent. It is provided in solid and solution formats for synthetic chemistry and assay development. Analytical documentation, including a certificate of analysis and safety data sheet, is available for lot-specific verification.
  • Used as a pharmaceutical intermediate and research reagent.
  • Available as solid and as 10 mM solution in DMSO.
  • High purity confirmed by COA (HPLC purity reported ≈99.95%).
  • Molecular formula C7H5ClO3 and molecular weight 172.57.
  • Safety and quality documentation (SDS, COA) available for download.
Catalog No. 50-002-99874