Learn More
Medchemexpress LLC 4-CMTB | 300851-67-6 | 99.7% | 294.80 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP1125
Encompass_Preferred
4-CMTB is a selective free fatty acid receptor 2 (FFA2/GPR43) agonist and a positive allosteric modulator (pEC50=6.38). It acts as both a direct agonist and a positive allosteric modulator of the action of short-chain free fatty acids by modulating the activity of short-chain fatty acids at FFA2 via the FFA2 second extracellular loop (ECL2). This product is for research use only.
  • Functions as a selective FFA2 agonist
  • Acts as a positive allosteric modulator
  • Modulates FFA2 activity via the FFA2 second extracellular loop (ECL2)
  • High solubility in DMSO at 100 mg/mL
  • Solid form storage at 4°C, protected from light
  • In solvent storage at -80°C for 6 months or -20°C for 1 month, protected from light
  • In vivo solubility options available for various formulations
Catalog No. 50-004-25470