Learn More
Medchemexpress LLC 4-cyano-N-cyclopropyl-N-(thiophen-2-ylmethyl)benzamide | 1210906-48-1 | 99.8% | 282.36 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY138283
Encompass_Preferred
MR837 is an inhibitor of NSD2-PWWP1, designed to bind with the PWWP domain of human nuclear receptor binding SET domain protein 2. This compound is intended for research use only.
  • Inhibits NSD2-PWWP1
  • Binds to human nuclear receptor binding SET domain protein 2 (PWWP domain)
  • High purity of 99.8%
  • Suitable for in vitro and in vivo studies with various solubility protocols
  • Stable in powder form for up to 3 years at -20°C
Catalog No. 50-003-64169