Learn More
Medchemexpress LLC 4'-Demethylepipodophyllotoxin | 6559-91-7 | 99.8% | 400.38 | 10 G

Supplier: Medchemexpress LLC HY1743510G
4'-Demethylepipodophyllotoxin is a laboratory chemical used in the manufacture of substances. This product is for research use only and has not been fully validated for medical applications.
- White to off-white solid
- High purity (99.76%)
- Molecular formula C21H20O8
- Molecular weight 400.38
- Suitable for laboratory research
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.